missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbutane-1,2,3-tricarboxylic Acid, TRC

Product Code. 30009763
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009763 10 g 10g
30007341 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009763 Supplier Toronto Research Chemicals Supplier No. TRCM29512010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77370-41-3
Molecular Formula C8H12O6
Molecular Weight (g/mol) 204.18
Synonym α,α-Dimethyltricarballylic Acid, 3-Methyl-1,2,3-butanecarboxylic Acid, 3-Methyl-1,2,3-butanetricarboxylic Acid, MBTCA
SMILES CC(C(C(O)=O)CC(O)=O)(C)C(O)=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H12O6/c1-8(2,7(13)14)4(6(11)12)3-5(9)10/h4H,3H2,1-2H3,(H,9,10)(H,11,12)(H,13,14)
Formula Weight 204.0634
Percent Purity >95%
Chemical Name or Material 3-Methylbutane-1,2,3-tricarboxylic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.