CAS RN 77370-41-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Methylbutane-1,2,3-tricarboxylic Acid, TRC
CAS: 77370-41-3 Molecular Formula: C8H12O6 Molecular Weight (g/mol): 204.18 Synonym: α,α-Dimethyltricarballylic Acid,3-Methyl-1,2,3-butanecarboxylic Acid,3-Methyl-1,2,3-butanetricarboxylic Acid,MBTCA SMILES: CC(C(C(O)=O)CC(O)=O)(C)C(O)=O