missing translation for 'onlineSavingsMsg'
Learn More

3'-Methylbiphenyl-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15465508
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465508 1 g 1g
15475508 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465508 Supplier Thermo Scientific Alfa Aesar Supplier No. H59257.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158619-46-6
Molecular Formula C14H11O2
Molecular Weight (g/mol) 211.24
MDL Number MFCD00558892
InChI Key BJVLFJADOVATRU-UHFFFAOYSA-M
Synonym 3'-methylbiphenyl-3-carboxylic acid, 3-3-methylphenyl benzoic acid, 3'-methyl 1,1'-biphenyl-3-carboxylic acid, 3'-methyl-1,1'-biphenyl-3-carboxylic acid, 3'-methyl-biphenyl-3-carboxylic acid, acmc-1ccsu, 3-m-tolyl benzoic acid, amtda074, cambridge id 5211690, 3-3-carboxyphenyl toluene
PubChem CID 747832
IUPAC Name 3-(3-methylphenyl)benzoic acid
SMILES CC1=CC=CC(=C1)C1=CC=CC(=C1)C([O-])=O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 212.25
Percent Purity 97%
Chemical Name or Material 3'-Methylbiphenyl-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.