accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 158619-46-6

IUPAC Name: 3'-methyl-[1,1'-biphenyl]-3-carboxylate
Synonyms: 3'-methyl-[1,1'-biphenyl]-3-carboxylate
Molecular Weight (g/mol): 211.24
Molecular Formula: C14H11O2
InChi Key: BJVLFJADOVATRU-UHFFFAOYSA-M
SMILES: CC1=CC=CC(=C1)C1=CC=CC(=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results