Learn More
3-Methoxythiophene, 99%
Product Code. 11465037
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465037 5 g 5g
11455037 1 g 1g
11475037 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11465037 Supplier Thermo Scientific Alfa Aesar Supplier No. B21940.06
Request Bulk or Custom Format
CAS: 17573-92-1 | C5H6OS | 114.162 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17573-92-1
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.162
MDL Number MFCD00043894
InChI Key RFSKGCVUDQRZSD-UHFFFAOYSA-N
Synonym thiophene, 3-methoxy, 3-methoxy-thiophene, pubchem8863, acmc-1bow6, 3-methoxythiophene, ksc491i4p, bidd:gt0411
PubChem CID 140249
IUPAC Name 3-methoxythiophene
SMILES COC1=CSC=C1
Specifications
Density 1.147
Boiling Point 60°C to 61°C (12 mmHg)
Flash Point 49°C (120°F)
Odor Recognizable
Refractive Index 1.532
Quantity 5 g
UN Number UN3271
Beilstein 106404
Sensitivity Air sensitive
Formula Weight 114.17
Percent Purity 99%
Chemical Name or Material 3-Methoxythiophene