CAS RN 17573-92-1
CAS RN 17573-92-1
IUPAC Name:
3-methoxythiophene
Synonyms:
p-methoxythiophene
Molecular Weight (g/mol):
114.16
Molecular Formula:
C5H6OS
InChi Key:
RFSKGCVUDQRZSD-UHFFFAOYSA-N
SMILES:
COC1=CSC=C1
1
–
2
of
2
results
1
3-Methoxythiophene, 99%
CAS: 17573-92-1 Molecular Formula: C5H6OS Molecular Weight (g/mol): 114.162 MDL Number: MFCD00043894 InChI Key: RFSKGCVUDQRZSD-UHFFFAOYSA-N Synonym: thiophene, 3-methoxy,3-methoxy-thiophene,pubchem8863,acmc-1bow6,3-methoxythiophene,ksc491i4p,bidd:gt0411 PubChem CID: 140249 IUPAC Name: 3-methoxythiophene SMILES: COC1=CSC=C1
2
3-Methoxythiophene, 98%
CAS: 17573-92-1 Molecular Formula: C5H6OS Molecular Weight (g/mol): 114.17 MDL Number: MFCD00043894 InChI Key: RFSKGCVUDQRZSD-UHFFFAOYSA-N Synonym: thiophene, 3-methoxy,3-methoxy-thiophene,pubchem8863,acmc-1bow6,3-methoxythiophene,ksc491i4p,bidd:gt0411 PubChem CID: 140249 IUPAC Name: 3-methoxythiophene SMILES: COC1=CSC=C1