Learn More
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Color | Brown |
| Density | 1.1430g/mL |
| Boiling Point | 80.0°C to 82.0°C (65.0 mmHg) |
| Flash Point | 49°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5300 to 1.5320 (20°C, 589nm) |
| Vedi altri risultati |
Sicurezza e movimentazione
- 3-Methoxythiophene
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.