Learn More
3-Isopropylcatechol, TRC
Product Code. 30398627
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398627 25 mg -
30396342 100 mg 100mg
30390515 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30398627 Supplier Toronto Research Chemicals Supplier No. TRCI82199525MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2138-48-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
Synonym 3-(1-Methylethyl)-1,2-benzenediol, 3-Isopropylpyrocatechol, 1,2-Dihydroxy-3-isopropylbenzene, 3-Isopropyl-1,2-dihydroxybenzene, 3-Isopropylbenzene-1,2-diol, NSC 62675
SMILES CC(C)c1cccc(O)c1O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C9H12O2/c1-6(2)7-4-3-5-8(10)9(7)11/h3-6,10-11H,1-2H3
Formula Weight 152.0837
Chemical Name or Material 3-Isopropylcatechol