CAS RN 2138-48-9
CAS RN 2138-48-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Isopropylcatechol, TRC
CAS: 2138-48-9 Molecular Formula: C9H12O2 Molecular Weight (g/mol): 152.19 Synonym: 3-(1-Methylethyl)-1,2-benzenediol,3-Isopropylpyrocatechol,1,2-Dihydroxy-3-isopropylbenzene,3-Isopropyl-1,2-dihydroxybenzene,3-Isopropylbenzene-1,2-diol,NSC 62675 SMILES: CC(C)c1cccc(O)c1O