missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Formylindole-1-acetic acid, 97+%

Product Code. p-7029353
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11386919 1 g 1g
11396919 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386919 Supplier Alfa Aesar™ Supplier No. H26807.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 138423-98-0
Molecular Formula C11H8NO3
Molecular Weight (g/mol) 202.19
MDL Number MFCD00452923
InChI Key ZUUGBTJTGRTIFK-UHFFFAOYSA-M
Synonym 3-formylindol-1-yl-acetic acid, 3-formylindol-1-yl acetic acid, 3-formyl-1-indolyl acetic acid, 3-formyl-indol-1-yl-acetic acid, 3-formyl-1h-indol-1-yl acetic acid, n-acetic acid-indole-3-carboxaldehyde, 2-3-formyl-1h-indol-1-yl acetic acid, 2-3-formylindol-1-yl acetic acid, 1h-indole-1-acetic acid, 3-formyl, indole-n-acetic acid-3-carboxaldehyde
PubChem CID 771465
SMILES [O-]C(=O)CN1C=C(C=O)C2=CC=CC=C12

Specifications

Melting Point ∽205°C (decomposition)
Quantity 1 g
Beilstein 5431204
Sensitivity Air Sensitive
IUPAC Name 2-(3-formyl-1H-indol-1-yl)acetate
Formula Weight 203.2
Percent Purity ≥97%
Chemical Name or Material 3-Formylindole-1-acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.