accessibility menu, dialog, popup

UserName

CAS RN 138423-98-0

OONO

CAS RN 138423-98-0

IUPAC Name: 2-(3-formyl-1H-indol-1-yl)acetate
Synonyms: 2-(3-formylindol-1-yl)acetate
Molecular Weight (g/mol): 202.19
Molecular Formula: C11H8NO3
InChi Key: ZUUGBTJTGRTIFK-UHFFFAOYSA-M
SMILES: [O-]C(=O)CN1C=C(C=O)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results