missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™

Product Code. 11373016
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373016 1 g 1g
11383016 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373016 Supplier Thermo Scientific Alfa Aesar Supplier No. L10836.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Fluoro-5-(trifluoromethyl)benzylamine may be employed as amine to generate different trityl-based pro-tags, by treating with activated carboxyl group-containing trityl synthons.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150517-77-4
Molecular Formula C8H7F4N
Molecular Weight (g/mol) 193.145
MDL Number MFCD00061163
InChI Key ADHUUMWSWNPEMZ-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethyl benzylamine, 3-fluoro-5-trifluoromethyl phenyl methanamine, 3-fluoro-5-trifluoromethylbenzylamine, 3-fluoro-5-trifluoromethyl-benzylamine, benzenemethanamine, 3-fluoro-5-trifluoromethyl, 1-3-fluoro-5-trifluoromethyl phenyl methanamine, 5-fluoro-3-trifluoromethyl phenyl methylamine, acmc-1c7bq, ksc495o0t
PubChem CID 2733304
IUPAC Name [3-fluoro-5-(trifluoromethyl)phenyl]methanamine
SMILES C1=C(C=C(C=C1C(F)(F)F)F)CN

Specifications

Density 1.319
Flash Point 76°C (168°F)
Refractive Index 1.451
Quantity 1 g
UN Number UN2735
Beilstein 7807166
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 193.15
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.