accessibility menu, dialog, popup

UserName

NH2FFFF

CAS RN 150517-77-4

IUPAC Name: 1-[3-fluoro-5-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[3-fluoro-5-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 193.15
Molecular Formula: C8H7F4N
InChi Key: ADHUUMWSWNPEMZ-UHFFFAOYSA-N
SMILES: NCC1=CC(F)=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results