missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(trifluoromethoxy)phenylacetic acid, 97%, Thermo Scientific™

Product Code. 15498085
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498085 250 mg 250mg
15488085 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498085 Supplier Thermo Scientific Alfa Aesar Supplier No. H32437.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017779-77-9
Molecular Formula C9H6ClF3O3
Molecular Weight (g/mol) 254.59
MDL Number MFCD09832392
InChI Key MYYLIFIHZIRCPZ-UHFFFAOYSA-N
Synonym 3-chloro-4-trifluoromethoxy phenylacetic acid, 2-3-chloro-4-trifluoromethoxy phenyl acetic acid, 3-chloro-4-trifluoromethoxy phenyl acetic acid, 3-chloro-4-trifluoromethoxy phenylacetic acid, jrd
PubChem CID 20112199
IUPAC Name 2-[3-chloro-4-(trifluoromethoxy)phenyl]acetic acid
SMILES OC(=O)CC1=CC(Cl)=C(OC(F)(F)F)C=C1

Specifications

Melting Point 72°C to 76°C
Quantity 250 mg
Formula Weight 254.59
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(trifluoromethoxy)phenylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.