accessibility menu, dialog, popup

UserName

OOHClOFFF

CAS RN 1017779-77-9

IUPAC Name: 2-[3-chloro-4-(trifluoromethoxy)phenyl]acetic acid
Synonyms: [3-chloro-4-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol): 254.59
Molecular Formula: C9H6ClF3O3
InChi Key: MYYLIFIHZIRCPZ-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(Cl)=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results