missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(trifluoromethoxy)benzylamine, 97%, Thermo Scientific™

Product Code. 15484417
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484417 250 mg 250mg
15474417 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484417 Supplier Thermo Scientific Alfa Aesar Supplier No. H31785.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
They are used as organic solvents and as flavoring agent.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771581-60-3
Molecular Formula C8H7ClF3NO
Molecular Weight (g/mol) 225.595
MDL Number MFCD06212980
InChI Key OVFDZNSGNUXERC-UHFFFAOYSA-N
Synonym 3-chloro-4-trifluoromethoxy phenyl methanamine, 1-3-chloro-4-trifluoromethoxy phenyl methanamine, 3-chloro-4-trifluoromethoxy benzylamine, 3-chloro-4-trifluoromethoxy benzylamine, jrd, 3-chloro-4-trifluoromethoxy phenyl methylamine, benzenemethanamine, 3-chloro-4-trifluoromethoxy
PubChem CID 53414455
IUPAC Name [3-chloro-4-(trifluoromethoxy)phenyl]methanamine
SMILES C1=CC(=C(C=C1CN)Cl)OC(F)(F)F

Specifications

Refractive Index 1.476
Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 225.6
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(trifluoromethoxy)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.