CAS RN 771581-60-3
IUPAC Name:
1-[3-chloro-4-(trifluoromethoxy)phenyl]methanamine
Synonyms:
1-[3-chloro-4-(trifluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol):
225.6
Molecular Formula:
C8H7ClF3NO
InChi Key:
OVFDZNSGNUXERC-UHFFFAOYSA-N
SMILES:
NCC1=CC(Cl)=C(OC(F)(F)F)C=C1
1
–
1
of
1
results
1
3-Chloro-4-(trifluoromethoxy)benzylamine, 97%, Thermo Scientific™
CAS: 771581-60-3 Molecular Formula: C8H7ClF3NO Molecular Weight (g/mol): 225.595 MDL Number: MFCD06212980 InChI Key: OVFDZNSGNUXERC-UHFFFAOYSA-N Synonym: 3-chloro-4-trifluoromethoxy phenyl methanamine,1-3-chloro-4-trifluoromethoxy phenyl methanamine,3-chloro-4-trifluoromethoxy benzylamine,3-chloro-4-trifluoromethoxy benzylamine, jrd,3-chloro-4-trifluoromethoxy phenyl methylamine,benzenemethanamine, 3-chloro-4-trifluoromethoxy PubChem CID: 53414455 IUPAC Name: [3-chloro-4-(trifluoromethoxy)phenyl]methanamine SMILES: C1=CC(=C(C=C1CN)Cl)OC(F)(F)F