missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,4,5,6-tetrafluoropyridine, 98%, Thermo Scientific™

Product Code. 11423517
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423517 5 g 5g
11433517 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423517 Supplier Thermo Scientific Alfa Aesar Supplier No. A17285.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1735-84-8
Molecular Formula C5ClF4N
Molecular Weight (g/mol) 185.51
MDL Number MFCD00010624
InChI Key UXUZMYHSICIOQT-UHFFFAOYSA-N
PubChem CID 137183
IUPAC Name 3-chloro-2,4,5,6-tetrafluoropyridine
SMILES FC1=NC(F)=C(Cl)C(F)=C1F

Specifications

Density 1.641
Boiling Point 118°C to 121°C
Flash Point >110°C (230°F)
Refractive Index 1.434
Quantity 5 g
UN Number UN1760
Beilstein 475873
Synonym pyridine, 3-chloro-2,4,5,6-tetrafluoro, 3-chlorotetrafluoropyridine, 2-chloro-2,4,5,6-tetrafluoropyridine, pubchem9226, 2,3,4,6-tetrafluoro-5-chloropyridine, 5-chloro-2,3,4,6-tetrafluoropyridine, 3-chloro-2,4,5,6-tetrafluoro-pyridine, pyridine,3-chloro-2,4,5,6-tetrafluoro, 3-chloro-2,4,5,6-tetrafluoropyridine, 3-chloranyl-2,4,5,6-tetrakis fluoranyl pyridine
Formula Weight 185.51
Percent Purity 98%
Chemical Name or Material 3-Chloro-2,4,5,6-tetrafluoropyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.