accessibility menu, dialog, popup

UserName

FNFClFF

CAS RN 1735-84-8

IUPAC Name: 3-chloro-2,4,5,6-tetrafluoropyridine
Synonyms: 3-chloro-2,4,5,6-tetrafluoropyridine
Molecular Weight (g/mol): 185.51
Molecular Formula: C5ClF4N
InChi Key: UXUZMYHSICIOQT-UHFFFAOYSA-N
SMILES: FC1=NC(F)=C(Cl)C(F)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results