Learn More
3-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™
Product Code. 15429025
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15429025 250 mg 250mg
15419025 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15429025 Supplier Thermo Scientific Alfa Aesar Supplier No. H35479.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 354548-73-5
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.04
MDL Number MFCD02182241
InChI Key UHBDEEITGHGHKK-UHFFFAOYSA-N
Synonym 3-bromoimidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylicacid, 3-bromo, 3-bromoimidazo 1,2-a pyridine-2-carboxylic acid hydrate, 3-bromoh-imidazo 1,2-a pyridine-2-carboxylic acid, 3-bromo-4-hydroimidazo 1,2-a pyridine-2-carboxylic acid
PubChem CID 16210784
IUPAC Name 3-bromoimidazo[1,2-a]pyridine-2-carboxylic acid
SMILES OC(=O)C1=C(Br)N2C=CC=CC2=N1
Specifications
Quantity 250 mg
Formula Weight 241.05 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 3-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid hydrate