accessibility menu, dialog, popup

UserName

NNBrOHO

CAS RN 354548-73-5

IUPAC Name: 3-bromoimidazo[1,2-a]pyridine-2-carboxylic acid
Synonyms: 3-bromoimidazo[1,2-a]pyridine-2-carboxylic acid
Molecular Weight (g/mol): 241.04
Molecular Formula: C8H5BrN2O2
InChi Key: UHBDEEITGHGHKK-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Br)N2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results