Learn More
3-Bromo-5-(trifluoromethoxy)benzamide, 97%, Thermo Scientific™
Product Code. 15485567
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485567 1 g 1g
15495567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15485567 Supplier Thermo Scientific Alfa Aesar Supplier No. H32805.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 914636-32-1
Molecular Formula C8H5BrF3NO2
Molecular Weight (g/mol) 284.032
MDL Number MFCD08458075
InChI Key PIHJYRPSTBFOOV-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethoxy benzamide
PubChem CID 26985420
IUPAC Name 3-bromo-5-(trifluoromethoxy)benzamide
SMILES C1=C(C=C(C=C1OC(F)(F)F)Br)C(=O)N
Specifications
Melting Point 144°C to 146°C
Quantity 1 g
Formula Weight 284.03
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(trifluoromethoxy)benzamide