accessibility menu, dialog, popup

UserName

CAS RN 914636-32-1

OFFFBrONH2

CAS RN 914636-32-1

IUPAC Name: 3-bromo-5-(trifluoromethoxy)benzamide
Synonyms: 3-bromo-5-(trifluoromethoxy)benzamide
Molecular Weight (g/mol): 284.03
Molecular Formula: C8H5BrF3NO2
InChi Key: PIHJYRPSTBFOOV-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(OC(F)(F)F)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results