missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluoro-5-iodobenzaldehyde, 97%, Thermo Scientific™

Product Code. 15466247
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466247 1 g 1g
15476247 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466247 Supplier Thermo Scientific Alfa Aesar Supplier No. H33441.03
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1356113-37-5
Molecular Formula C7H3BrFIO
Molecular Weight (g/mol) 328.91
MDL Number MFCD20486165
InChI Key VZRIOLAMLBZGDB-UHFFFAOYSA-N
PubChem CID 95567744
IUPAC Name 3-bromo-4-fluoro-5-iodobenzaldehyde
SMILES FC1=C(Br)C=C(C=O)C=C1I

Specifications

Quantity 1 g
Formula Weight 328.9
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-fluoro-5-iodobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.