CAS RN 1356113-37-5
CAS RN 1356113-37-5
IUPAC Name:
3-bromo-4-fluoro-5-iodobenzaldehyde
Synonyms:
3-bromo-4-fluoro-5-iodobenzaldehyde
Molecular Weight (g/mol):
328.91
Molecular Formula:
C7H3BrFIO
InChi Key:
VZRIOLAMLBZGDB-UHFFFAOYSA-N
SMILES:
FC1=C(Br)C=C(C=O)C=C1I
1
–
1
of
1
results
1
3-Bromo-4-fluoro-5-iodobenzaldehyde, 97%, Thermo Scientific™
CAS: 1356113-37-5 Molecular Formula: C7H3BrFIO Molecular Weight (g/mol): 328.91 MDL Number: MFCD20486165 InChI Key: VZRIOLAMLBZGDB-UHFFFAOYSA-N PubChem CID: 95567744 IUPAC Name: 3-bromo-4-fluoro-5-iodobenzaldehyde SMILES: FC1=C(Br)C=C(C=O)C=C1I