missing translation for 'onlineSavingsMsg'
Learn More

(3-Benzoylphenyl)acetonitrile, TRC

Product Code. 17921145
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17921145 250 mg 250mg
17914645 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17921145 Supplier Toronto Research Chemicals Supplier No. TRCB208350250MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21288-34-6
Molecular Formula C15 H11 N O
Molecular Weight (g/mol) 221.25
Synonym (3-Benzoylphenyl)ethanenitrile, Ketoprofen Imp. I (EP)
IUPAC Name 2-(3-benzoylphenyl)acetonitrile
SMILES O=C(c1ccccc1)c2cccc(CC#N)c2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H11NO/c16-10-9-12-5-4-8-14(11-12)15(17)13-6-2-1-3-7-13/h1-8,11H,9H2
Formula Weight 221.0841
Chemical Name or Material (3-Benzoylphenyl)ethanenitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.