CAS RN 21288-34-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(3-Benzoylphenyl)acetonitrile, TRC
CAS: 21288-34-6 Molecular Formula: C15 H11 N O Molecular Weight (g/mol): 221.25 Synonym: (3-Benzoylphenyl)ethanenitrile,Ketoprofen Imp. I (EP) IUPAC Name: 2-(3-benzoylphenyl)acetonitrile SMILES: O=C(c1ccccc1)c2cccc(CC#N)c2
2
(3-Benzoylphenyl)ethanenitrile, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.