Learn More
3-Amino-5-cyclobutyl-1H-pyrazole, 97%
Product Code. p-7033723
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479570 250 mg 250mg
11489570 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11479570 Supplier Thermo Scientific Alfa Aesar Supplier No. H50880.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 326827-21-8 | C7H11N3 | 137.19 g/mol
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 326827-21-8
Molecular Formula C7H11N3
Molecular Weight (g/mol) 137.19
MDL Number MFCD08061045,MFCD11187387
InChI Key DELFRVWPWUEOHU-UHFFFAOYSA-N
Synonym 3-amino-5-cyclobutyl-1h-pyrazole, 3-cyclobutyl-1h-pyrazol-5-amine, 1h-pyrazol-3-amine, 5-cyclobutyl, 5-cyclobutyl-1h-pyrazol-3-ylamine, 5-cyclobutyl-2h-pyrazol-3-amine, 5-cyclobutyl-2h-pyrazol-3-ylamine, 3-amino-5-cyclobutyl-2h-pyrazole, acmc-20agky, 5-amino-3-cyclobutylpyrazole
PubChem CID 22176774
IUPAC Name 5-cyclobutyl-1H-pyrazol-3-amine
SMILES NC1=NNC(=C1)C1CCC1
Specifications
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 137.19
Percent Purity 97%
Chemical Name or Material 3-Amino-5-cyclobutyl-1H-pyrazole