accessibility menu, dialog, popup

UserName

NNHH2N

CAS RN 326827-21-8

IUPAC Name: 5-cyclobutyl-1H-pyrazol-3-amine
Synonyms: 5-cyclobutyl-1H-pyrazol-3-amine
Molecular Weight (g/mol): 137.19
Molecular Formula: C7H11N3
InChi Key: DELFRVWPWUEOHU-UHFFFAOYSA-N
SMILES: NC1=NNC(=C1)C1CCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results