missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-L-tyrosine Hydrochloride, TRC

Product Code. 30136439
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136439 2.5 g 2.5g
30136812 250 mg 250mg
30012873 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136439 Supplier Toronto Research Chemicals Supplier No. TRCA1897502.5G
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32404-28-7
Molecular Formula C11H14ClNO4
Molecular Weight (g/mol) 259.69
IUPAC Name (2S)-3-(3-acetyl-4-hydroxyphenyl)-2-aminopropanoic acid
SMILES N[C@H](C(O)=O)CC1=CC(C(C)=O)=C(O)C=C1.Cl

Specifications

InChI Formula InChI=1S/C11H13NO4/c1-6(13)8-4-7(2-3-10(8)14)5-9(12)11(15)16/h2-4,9,14H,5,12H2,1H3,(H,15,16)/t9-/m0/s1
Quantity 2.5 g
Formula Weight 259.0611
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material 3-Acetyl-L-tyrosine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.