CAS RN 32404-28-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Acetyl-L-tyrosine Hydrochloride, TRC
CAS: 32404-28-7 Molecular Formula: C11H14ClNO4 Molecular Weight (g/mol): 259.69 IUPAC Name: (2S)-3-(3-acetyl-4-hydroxyphenyl)-2-aminopropanoic acid SMILES: N[C@H](C(O)=O)CC1=CC(C(C)=O)=C(O)C=C1.Cl