missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane, 95%

Product Code. p-7046258
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373197 250 mg 250mg
11353197 1 g 1g
11363197 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11373197 Supplier Thermo Scientific Alfa Aesar Supplier No. L13709.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 732306-23-9 | C10H9ClF6Si | 306.707 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 732306-23-9
Molecular Formula C10H9ClF6Si
Molecular Weight (g/mol) 306.707
MDL Number MFCD01862186
InChI Key JZWDIKPZWMXPMG-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenyldimethylchlorosilane, 3,5-bis trifluoromethyl phenyl chloro dimethylsilane, 3,5-bis trifluoromethyl phenyl chloro dimethyl silane, 3,5-bis trifluoromethyl phenyl-chloro-dimethylsilane, 3,5-bis trifluoromethyl phenyl dimethylsilyl chloride, benzene,1-chlorodimethylsilyl-3,5-bis trifluoromethyl
PubChem CID 2783240
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]-chloro-dimethylsilane
SMILES C[Si](C)(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)Cl

Specifications

Boiling Point 95°C to 96°C (30mmHg)
Quantity 250 mg
UN Number UN3265
Formula Weight 306.71
Percent Purity 95%
Refractive Index 1.424
Sensitivity Moisture sensitive
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.