accessibility menu, dialog, popup

UserName

CH3SiH3CFFFFFFCl

CAS RN 732306-23-9

IUPAC Name: [3,5-bis(trifluoromethyl)phenyl](chloro)dimethylsilane
Synonyms: [3,5-bis(trifluoromethyl)phenyl](chloro)dimethylsilane
Molecular Weight (g/mol): 306.71
Molecular Formula: C10H9ClF6Si
InChi Key: JZWDIKPZWMXPMG-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results