missing translation for 'onlineSavingsMsg'
Learn More

3-[(4-Formylphenoxy)methyl]thiophene-2-carbonitrile, Thermo Scientific™

Product Code. 11385539
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11385539 1 g 1g
11375539 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11385539 Supplier Thermo Scientific Alfa Aesar Supplier No. H50479.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 256471-05-3
Molecular Formula C13H9NO2S
Molecular Weight (g/mol) 243.28
MDL Number MFCD00115160
InChI Key REEBWSYYNPPSKV-UHFFFAOYSA-N
Synonym 3-4-formylphenoxy methyl thiophene-2-carbonitrile, 3-4-formylphenoxymethyl thiophene-2-carbonitrile, acmc-1cou6
PubChem CID 2797328
IUPAC Name 3-[(4-formylphenoxy)methyl]thiophene-2-carbonitrile
SMILES O=CC1=CC=C(OCC2=C(SC=C2)C#N)C=C1

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 243.29
Chemical Name or Material 3-[(4-Formylphenoxy)methyl]thiophene-2-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.