accessibility menu, dialog, popup

UserName

CAS RN 256471-05-3

OOSN

CAS RN 256471-05-3

IUPAC Name: 3-[(4-formylphenoxy)methyl]thiophene-2-carbonitrile
Synonyms: 3-(4-formylphenoxymethyl)thiophene-2-carbonitrile
Molecular Weight (g/mol): 243.28
Molecular Formula: C13H9NO2S
InChi Key: REEBWSYYNPPSKV-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(OCC2=C(SC=C2)C#N)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results