missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxythiophenol, 98%, Thermo Scientific™

Product Code. 11437796
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437796 5 g 5g
11427796 1 g 1g
11447796 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437796 Supplier Thermo Scientific Alfa Aesar Supplier No. A14745.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-96-9
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.226
MDL Number MFCD00052823
InChI Key MTKAJLNGIVXZIS-UHFFFAOYSA-N
Synonym 3,4-dimethoxythiophenol, 3,4-dimethoxy thiophenol, 3,4-dimethoxybenzene-1-thiol, 3,4-dimethoxy-thiophenol, benzenethiol, 3,4-dimethoxy, 4-mercaptoveratrole, pubchem6823, acmc-1bdpf, 3,4-dimethoxy-benzenethiol, 3,4-dimethoxyphenylmercaptan
PubChem CID 136539
IUPAC Name 3,4-dimethoxybenzenethiol
SMILES COC1=C(C=C(C=C1)S)OC

Specifications

Density 1.19
Boiling Point 263°C
Flash Point >113°C (235°F)
Odor Stench
Refractive Index 1.59
Quantity 5 g
Beilstein 1945136
Sensitivity Air Sensitive
Formula Weight 170.23
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxythiophenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.