CAS RN 700-96-9
IUPAC Name:
3,4-dimethoxybenzene-1-thiol
Synonyms:
3,4-dimethoxybenzenethiol
Molecular Weight (g/mol):
170.23
Molecular Formula:
C8H10O2S
InChi Key:
MTKAJLNGIVXZIS-UHFFFAOYSA-N
SMILES:
COC1=CC=C(S)C=C1OC
1
–
1
of
1
results
1
3,4-Dimethoxythiophenol, 98%, Thermo Scientific™
CAS: 700-96-9 Molecular Formula: C8H10O2S Molecular Weight (g/mol): 170.226 MDL Number: MFCD00052823 InChI Key: MTKAJLNGIVXZIS-UHFFFAOYSA-N Synonym: 3,4-dimethoxythiophenol,3,4-dimethoxy thiophenol,3,4-dimethoxybenzene-1-thiol,3,4-dimethoxy-thiophenol,benzenethiol, 3,4-dimethoxy,4-mercaptoveratrole,pubchem6823,acmc-1bdpf,3,4-dimethoxy-benzenethiol,3,4-dimethoxyphenylmercaptan PubChem CID: 136539 IUPAC Name: 3,4-dimethoxybenzenethiol SMILES: COC1=C(C=C(C=C1)S)OC