missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethoxyflavone, 97%, Thermo Scientific™

Product Code. p-7046391
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369015 100 mg 100mg
11359015 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369015 Supplier Thermo Scientific Alfa Aesar Supplier No. L14160.MC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4143-62-8
Molecular Formula C17H14O4
Molecular Weight (g/mol) 282.295
MDL Number MFCD00143009
InChI Key ZGHORMOOTZTQFL-UHFFFAOYSA-N
Synonym 3',4'-dimethoxyflavone, 2-3,4-dimethoxyphenyl-4h-chromen-4-one, 2-3,4-dimethoxyphenyl chromen-4-one, chembl91153, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl, 3',4'-dmf, pubchem9847, spectrum_001039, acmc-1apwv, specplus_000555
PubChem CID 688674
IUPAC Name 2-(3,4-dimethoxyphenyl)chromen-4-one
SMILES COC1=C(C=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2)OC

Specifications

Melting Point 154°C to 155°C
Quantity 100 mg
Beilstein 236749
Formula Weight 282.3
Percent Purity 97%
Chemical Name or Material 3',4'-Dimethoxyflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.