accessibility menu, dialog, popup

UserName

CH3OOH3COO

CAS RN 4143-62-8

IUPAC Name: 2-(3,4-dimethoxyphenyl)-4H-chromen-4-one
Synonyms: 2-(3,4-dimethoxyphenyl)chromen-4-one
Molecular Weight (g/mol): 282.3
Molecular Formula: C17H14O4
InChi Key: ZGHORMOOTZTQFL-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)C1=CC(=O)C2=CC=CC=C2O1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results