missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzyl alcohol, 99%, Thermo Scientific™

Product Code. 11439196
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439196 5 g 5g
11429196 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439196 Supplier Thermo Scientific Alfa Aesar Supplier No. A18051.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85118-05-4
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.12
MDL Number MFCD00010628
InChI Key GNQLTCVBSGVGHC-UHFFFAOYSA-N
PubChem CID 522833
IUPAC Name (3,4-difluorophenyl)methanol
SMILES OCC1=CC=C(F)C(F)=C1

Specifications

Density 1.262
Flash Point 97°C (206°F)
Refractive Index 1.49
Quantity 5 g
Beilstein 9043426
Synonym 3,4-difluorobenzyl alcohol, 3,4-difluorophenyl methanol, benzenemethanol, 3,4-difluoro, 3,4-difluorobenzylalcohol, 3,4-difluoro-phenyl-methanol, 3,4-difluorophenyl methan-1-ol, 3, 4-difluorobenzyl alcohol, pubchem4916, acmc-209q4o, ksc493q4f
Formula Weight 144.12
Percent Purity 99%
Chemical Name or Material 3,4-Difluorobenzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.