accessibility menu, dialog, popup

UserName

CAS RN 85118-05-4

IUPAC Name: (3,4-difluorophenyl)methanol
Synonyms: (3,4-difluorophenyl)methanol
Molecular Weight (g/mol): 144.12
Molecular Formula: C7H6F2O
InChi Key: GNQLTCVBSGVGHC-UHFFFAOYSA-N
SMILES: OCC1=CC=C(F)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results