Learn More
3,3,3-Trifluoro-2-(trifluoromethyl)propionyl chloride, TRC
Product Code. 30453882
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453882 2.5 g 2.5g
30449979 250 mg 250mg
30451407 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30453882 Supplier Toronto Research Chemicals Supplier No. TRCT9001152.5G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 382-19-4
Molecular Formula C4 H Cl F6 O
Molecular Weight (g/mol) 214.49
IUPAC Name 3,3,3-trifluoro-2-(trifluoromethyl)propanoyl chloride
SMILES FC(F)(F)C(C(=O)Cl)C(F)(F)F
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C4HClF6O/c5-2(12)1(3(6,7)8)4(9,10)11/h1H
Formula Weight 213.962
Chemical Name or Material 3,3,3-Trifluoro-2-(trifluoromethyl)propionyl chloride