CAS RN 382-19-4
CAS RN 382-19-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,3,3-Trifluoro-2-(trifluoromethyl)propionyl chloride, TRC
CAS: 382-19-4 Molecular Formula: C4 H Cl F6 O Molecular Weight (g/mol): 214.49 IUPAC Name: 3,3,3-trifluoro-2-(trifluoromethyl)propanoyl chloride SMILES: FC(F)(F)C(C(=O)Cl)C(F)(F)F