Learn More
2-Propylfuran, TRC
Product Code. 30420997
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420997 50 mg -
30410375 100 mg -
30421892 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30420997 Supplier Toronto Research Chemicals Supplier No. TRCP83937350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4229-91-8
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.15
IUPAC Name 2-propylfuran
SMILES CCCc1occc1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C7H10O/c1-2-4-7-5-3-6-8-7/h3,5-6H,2,4H2,1H3
Formula Weight 110.0732
Percent Purity >95%
Chemical Name or Material 2-Propylfuran
Grade GC