CAS RN 4229-91-8
CAS RN 4229-91-8
IUPAC Name:
2-propylfuran
Synonyms:
furan, 2-propyl-
Molecular Weight (g/mol):
110.16
Molecular Formula:
C7H10O
InChi Key:
CPLJMYOQYRCCBY-UHFFFAOYSA-N
SMILES:
CCCC1=CC=CO1
1
–
2
of
2
results
1
2-Propylfuran, TRC
CAS: 4229-91-8 Molecular Formula: C7H10O Molecular Weight (g/mol): 110.15 IUPAC Name: 2-propylfuran SMILES: CCCc1occc1
2
2-n-Propylfuran, 99%
CAS: 4229-91-8 Molecular Formula: C7H10O Molecular Weight (g/mol): 110.16 MDL Number: MFCD00047070 InChI Key: CPLJMYOQYRCCBY-UHFFFAOYSA-N Synonym: furan, 2-propyl,2-n-propylfuran,2-n-propyl furan,furan, propyl,2-propyl-furan,furan, alpha-propyl,acmc-1ctud,2-propyl furan,furan, .alpha.-propyl,cpljmyoqyrccby-uhfffaoysa PubChem CID: 77907 IUPAC Name: 2-propylfuran SMILES: CCCC1=CC=CO1