missing translation for 'onlineSavingsMsg'
Learn More

2'-Phenoxymethanesulfonanilide, TRC

Product Code. 30422973
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422973 25 g 25g
30411592 100 mg -
30411918 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30422973 Supplier Toronto Research Chemicals Supplier No. TRCP31861825G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51765-51-6
Molecular Formula C13 H13 N O3 S
Molecular Weight (g/mol) 263.31
Synonym N-(2-Phenoxyphenyl)methanesulphonamide, Nimesulide Imp. B (EP)
IUPAC Name N-(2-phenoxyphenyl)methanesulfonamide
SMILES CS(=O)(=O)Nc1ccccc1Oc2ccccc2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C13H13NO3S/c1-18(15,16)14-12-9-5-6-10-13(12)17-11-7-3-2-4-8-11/h2-10,14H,1H3
Formula Weight 263.0616
Chemical Name or Material N-(2-Phenoxyphenyl)methanesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.