CAS RN 51765-51-6
CAS RN 51765-51-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2'-Phenoxymethanesulfonanilide, TRC
CAS: 51765-51-6 Molecular Formula: C13 H13 N O3 S Molecular Weight (g/mol): 263.31 Synonym: N-(2-Phenoxyphenyl)methanesulphonamide,Nimesulide Imp. B (EP) IUPAC Name: N-(2-phenoxyphenyl)methanesulfonamide SMILES: CS(=O)(=O)Nc1ccccc1Oc2ccccc2
2
N-(2-Phenoxyphenyl)methanesulphonamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.