Learn More
2-n-Pentylthiophene, 98%, Thermo Scientific™
Product Code. 11464947
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464947 5 g 5g
11474947 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11464947 Supplier Thermo Scientific Alfa Aesar Supplier No. B21885.06
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also used as flavoring agent for food.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4861-58-9
Molecular Formula C9H14S
Molecular Weight (g/mol) 154.271
MDL Number MFCD00041017
InChI Key NOYVOSGVFSEKPR-UHFFFAOYSA-N
Synonym 2-n-pentylthiophene, thiophene, 2-pentyl, 2-n-amylthiophene, 1-methylbutylthiophene, unii-l79los1zkm, l79los1zkm, 2-amylthiophene, 2-pentyl thiophene, 2-pentyl-thiophene, acmc-209kdh
PubChem CID 20995
IUPAC Name 2-pentylthiophene
SMILES CCCCCC1=CC=CS1
Specifications
Density 0.944
Boiling Point 198°C
Flash Point 75°C (167°F)
Refractive Index 1.4995
Quantity 5 g
Beilstein 107941
Solubility Information Slightly soluble in water.
Formula Weight 154.28
Percent Purity 98%
Chemical Name or Material 2-n-Pentylthiophene