accessibility menu, dialog, popup

UserName

H3CS

CAS RN 4861-58-9

IUPAC Name: 2-pentylthiophene
Synonyms: 2-pentylthiophene
Molecular Weight (g/mol): 154.27
Molecular Formula: C9H14S
InChi Key: NOYVOSGVFSEKPR-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results