Learn More
2-Methylcitric Acid, TRC
Product Code. 30010343
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010343 250 mg 250mg
30007630 500 mg 500mg
30003431 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010343 Supplier Toronto Research Chemicals Supplier No. TRCM265080250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6061-96-7
Molecular Formula C7 H10 O7
Molecular Weight (g/mol) 206.15
Synonym 1,2,3-Butanetricarboxylic acid, 2-hydroxy- (7CI, 8CI, 9CI, ACI), 2-Hydroxy-1,2,3-butanetricarboxylic acid (ACI), 2-Hydroxy-1-methylpropane-1,2,3-tricarboxylic acid, 2-Methylcitric acid
IUPAC Name 2-hydroxybutane-1,2,3-tricarboxylic acid
SMILES CC(C(=O)O)C(O)(CC(=O)O)C(=O)O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C7H10O7/c1-3(5(10)11)7(14,6(12)13)2-4(8)9/h3,14H,2H2,1H3,(H,8,9)(H,10,11)(H,12,13)
Formula Weight 206.0427
Percent Purity >95%
Chemical Name or Material 2-Methylcitric Acid
Grade HPLC